Merck
  • Diet and psoriasis, part II: celiac disease and role of a gluten-free diet.
Diet and psoriasis, part II: celiac disease and role of a gluten-free diet.
Journal of the American Academy of Dermatology (2014-05-02)
Bhavnit K Bhatia, Jillian W Millsop, Maya Debbaneh, John Koo, Eleni Linos, Wilson Liao
ABSTRACT
Patients with psoriasis have been shown to have a higher prevalence of other autoimmune diseases including celiac disease, a condition marked by sensitivity to dietary gluten. A number of studies suggest that psoriasis and celiac disease share common genetic and inflammatory pathways. Here we review the epidemiologic association between psoriasis and celiac disease and perform a meta-analysis to determine whether patients with psoriasis more frequently harbor serologic markers of celiac disease. We also examine whether a gluten-free diet can improve psoriatic skin disease.
Sigma-Aldrich
Gliadin from wheat
Sigma-Aldrich
Transglutaminase from guinea pig liver, lyophilized powder, ≥1.5 units/mg protein