Merck
  • Cerebrospinal fluid asparagine depletion during pegylated asparaginase therapy in children with acute lymphoblastic leukaemia.
Cerebrospinal fluid asparagine depletion during pegylated asparaginase therapy in children with acute lymphoblastic leukaemia.
British journal of haematology (2014-04-08)
Louise T Henriksen, Jacob Nersting, Raheel A Raja, Thomas L Frandsen, Steen Rosthøj, Henrik Schrøder, Birgitte K Albertsen
ABSTRACT
L-asparaginase is an important drug in the treatment of childhood acute lymphoblastic leukaemia (ALL). Cerebrospinal fluid (CSF) asparagine depletion is considered a marker of asparaginase effect in the central nervous system (CNS) and may play a role in CNS-directed anti-leukaemia therapy. The objective of this study was to describe CSF asparagine depletion during 30 weeks of pegylated asparaginase therapy, 1000 iu/m(2) i.m. every second week, and to correlate CSF asparagine concentration with serum L-asparaginase enzyme activity. Danish children (1-17 years) with ALL, treated according to the Nordic Society of Paediatric Haematology and Oncology ALL2008 protocol, standard and intermediate risk, were included. CSF samples were obtained throughout L-asparaginase treatment at every scheduled lumbar puncture. A total of 128 samples from 31 patients were available for analysis. Median CSF asparagine concentration decreased from a pre-treatment level of 5·3 μmol/l to median levels ≤1·5 μmol/l. However, only 4/31 patients (five samples) had CSF asparagine concentrations below the limit of detection (0·1 μmol/l). In 11 patients, 24 paired same day serum and CSF samples were obtained. A decrease in CSF asparagine corresponded to serum enzyme activities above 50 iu/l. Higher serum enzyme activities were not followed by more extensive depletion. In conclusion, pegylated asparaginase 1000 iu/m(2) i.m. every second week effectively reduced CSF asparagine levels.
Product Description
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Asparagine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture