Merck
  • Efficient solid-phase synthesis of 2-substituted-3-hydroxy-4(1H)-quinolinone-7-carboxamides with two diversity positions.
Efficient solid-phase synthesis of 2-substituted-3-hydroxy-4(1H)-quinolinone-7-carboxamides with two diversity positions.
Journal of combinatorial chemistry (2007-08-07)
Miroslav Soural, Viktor Krchnák
ABSTRACT
A highly efficient solid-phase synthesis of 2-substituted-3-hydroxy-4(1H)-quinolinone-7-carboxamides was developed using anthranilates and bromoketones as the key synthons. Primary amines immobilized to an acid-cleavable BAL linker were acylated with 1-methyl-2-aminoterephtalate. After cleavage of the methyl ester, bromoketones were used to form resin-bound phenacyl esters. Acid-mediated cleavage and subsequent cyclization in solution afforded 3-hydroxy-4(1H)-quinolinones in high purity and yield. Highly efficient solid-phase synthesis (purity>90%, yield>80%, synthetic time=2 days using commercially available synthons) is amenable to high-throughput/combinatorial synthesis to match the high-throughput screening capability.
Sigma-Aldrich
2-Bromo-4′-methylacetophenone, 90%