Merck
  • Cold-inducible RNA-binding protein mediates neuroinflammation in cerebral ischemia.
Cold-inducible RNA-binding protein mediates neuroinflammation in cerebral ischemia.
Biochimica et biophysica acta (2014-03-13)
Mian Zhou, Weng-Lang Yang, Youxin Ji, Xiaoling Qiang, Ping Wang
ABSTRACT
Neuroinflammation is a key cascade after cerebral ischemia. Excessive production of proinflammatory mediators in ischemia exacerbates brain injury. Cold-inducible RNA-binding protein (CIRP) is a newly discovered proinflammatory mediator that can be released into the circulation during hemorrhage or septic shock. Here, we examine the involvement of CIRP in brain injury during ischemic stroke. Stroke was induced by middle cerebral artery occlusion (MCAO). In vitro hypoxia was conducted in a hypoxia chamber containing 1% oxygen. CIRP and tumor necrosis factor-α (TNF-α) levels were assessed by RT-PCR and Western blot analysis. CIRP is elevated along with an upregulation of TNF-α expression in mouse brain after MCAO. In CIRP-deficient mice, the brain infarct volume, induction of TNF-α, and activation of microglia are markedly reduced after MCAO. Using microglial BV2 cells, we demonstrate that hypoxia induces the expression, translocation, and release of CIRP, which is associated with an increase of TNF-α levels. Addition of recombinant murine (rm) CIRP directly induces TNF-α release from BV2 cells and such induction is inhibited by neutralizing antisera to CIRP. Moreover, rmCIRP activates the NF-κB signaling pathway in BV2 cells. The conditioned medium from BV2 cells exposed to hypoxia triggers the apoptotic cascade by increasing caspase activity and decreasing Bcl-2 expression in neural SH-SY5Y cells, which is inhibited by antisera to CIRP. Extracellular CIRP is a detrimental factor in stimulating inflammation to cause neuronal damage in cerebral ischemia. Development of an anti-CIRP therapy may benefit patients with brain ischemia.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
SAFC
BIS-TRIS
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%