Merck
  • Stereoselective total synthesis of ieodomycins A and B and revision of the NMR spectroscopic data of ieodomycin B.
Stereoselective total synthesis of ieodomycins A and B and revision of the NMR spectroscopic data of ieodomycin B.
The Journal of organic chemistry (2013-06-21)
Sayantan Das, Rajib Kumar Goswami
ABSTRACT
Stereoselective total synthesis of antimicrobial marine metabolites ieodomycin A and B have been accomplished starting from commercially available 4-pentyn-1-ol featuring strategic application of the Negishi reaction, Kumada coupling, and Crimmins acetate aldol. This revises the proton NMR spectra of ieodomycin B.
Sigma-Aldrich
4-Pentyn-1-ol, 97%