Merck
  • Opioid δ₁ receptor antagonist 7-benzylidenenaltrexone as an effective resistance reverser for chloroquine-resistant Plasmodium chabaudi.
Opioid δ₁ receptor antagonist 7-benzylidenenaltrexone as an effective resistance reverser for chloroquine-resistant Plasmodium chabaudi.
Bioorganic & medicinal chemistry letters (2011-07-19)
Yoshinori Miyata, Hideaki Fujii, Yumiko Osa, Seiki Kobayashi, Tsutomu Takeuchi, Hiroshi Nagase
ABSTRACT
We evaluated antimalarial and/or chloroquine-resistance reversing effects of five opioid receptor antagonists. Although none of the evaluated compounds showed antimalarial effects, some of them, especially the δ(1) receptor antagonist, 7-benzylidenenaltrexone (BNTX) exhibited potent chloroquine-resistance reversing effects in Plasmodium chabaudi.
Sigma-Aldrich
BNTX maleate salt hydrate, ≥98% (HPLC)