Merck
  • Long-term effects of Helicobacter pylori infection on acid and pepsin secretion.
Long-term effects of Helicobacter pylori infection on acid and pepsin secretion.
The Yale journal of biology and medicine (1997-01-01)
F Halter, R K Zetterman
ABSTRACT
Chronic Helicobacter pylori infection causes a slight postprandial hypergastinemia, generally referred to as exaggerated or inappropriate gastrin release. This can be ablated by eradication of this infective agent. The expectations that this would further unravel the mysteries of the pathogenesis of peptic ulcer disease have not been fulfilled. It is now well established that of conventional acid secretory patterns such as basal acid secretion, maximum gastrin-stimulated acid secretion, and of sensitivity of the parietal cell to gastrin, only basal acid is modified by chronic H. pylori colonization. This particularly relates to basal secretion in duodenal ulcer patients, as basal secretion of otherwise healthy, chronically H. pylori-infected subjects appears to be affected in only a small proportion of subjects. It is of particular interest, however, that chronic H. pylori infection supplies a solid explanation why acid inhibitory pathways are deficient in duodenal ulcer disease, since this is reversible following H. pylori eradication as demonstrated by elegant studies with gastrin-releasing, peptide-stimulated acid secretion. Furthermore, it has gradually become apparent that exaggerated gastrin response is probably no more than an innocent bystander of chronic H. pylori infection. Paradoxically, in a small subset of patients, hypo-or anacidity accompanying chronic H. pylori infection can be reverted by H. pylori eradication, for currently unknown reasons.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥3200 units/mg protein