Merck
  • [Intracystic infusion therapy of minocycline hydrochloride for benign hepatic cysts].
[Intracystic infusion therapy of minocycline hydrochloride for benign hepatic cysts].
Nihon Ronen Igakkai zasshi. Japanese journal of geriatrics (1995-01-01)
M Ohhira, M Yamada, Y Murazumi, M Ohhira, Y Takasugi, M Namiki, T Suzuki
ABSTRACT
This paper describes ultrasonically-guided infusion of minocycline hydrochloride solution (MINO infusion therapy) in benign nonparasitic cysts of the liver. MINO infusion therapy was performed in 7 large hepatic cysts and successful results were obtained in 6 lesions. The infusion procedure in the initial 4 hepatic cysts a 7 Fr catheter was placed into the cyst and MINO solution was left in the cyst according to the procedure of ethanol infusion therapy. In the most recent 2 cases the cyst was punctured with a 21G needle, washed with physiologic saline and then infused MINO solution without placement of an indwelling drainage tube. This modified procedure is simple and safe and also yields a good therapeutic result. MINO infusion therapy for benign hepatic cyst is very useful and the modified procedure can be performed safely in elderly patients and in patients with several complications.
Sigma-Aldrich
Minocycline hydrochloride, powder