Merck
  • Intraocular sustained-release delivery systems for triamcinolone acetonide.
Intraocular sustained-release delivery systems for triamcinolone acetonide.
Pharmaceutical research (2009-02-03)
Saffar Mansoor, Baruch D Kuppermann, M Cristina Kenney
ABSTRACT
Recently, the use of triamcinolone acetonide (TA) injection has increased dramatically in treatment for several ocular diseases. Among them, macular diseases such as macular edema due to diabetic retinopathy, venous occlusive diseases, ocular inflammation and age-related macular degeneration (AMD) are very common vision threatening disorders and are great challenges to treat. In these types of chronic retinal diseases, repeated intraocular injections of TA are often required which increases the likelihood of complications. In order to achieve sustained-release, maintain therapeutic levels of TA over longer times and reduce frequency of intravitreal injections, researchers are investigating different implantable devices or injectable systems. However, as of yet, there is no sustained-release product for TA available on the commercial market. This review discusses and compares different sustained-release devices or injectable systems that are currently being developed.
Product Description
Triamcinolone acetonide, European Pharmacopoeia (EP) Reference Standard
Triamcinolone acetonide for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Triamcinolone acetonide, United States Pharmacopeia (USP) Reference Standard
Supelco
Triamcinolone acetonide, analytical standard