Merck
  • Bis-histidine-coordinated hemes in four-helix bundles: how the geometry of the bundle controls the axial imidazole plane orientations in transmembrane cytochromes of mitochondrial complexes II and III and related proteins.
Bis-histidine-coordinated hemes in four-helix bundles: how the geometry of the bundle controls the axial imidazole plane orientations in transmembrane cytochromes of mitochondrial complexes II and III and related proteins.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2008-04-18)
Edward A Berry, F Ann Walker
ABSTRACT
Early investigation of the electron paramagnetic resonance spectra of bis-histidine-coordinated membrane-bound ferriheme proteins led to the description of a spectral signal that had only one resolved feature. These became known as "highly anisotropic low-spin" or "large g(max)" ferriheme centers. Extensive work with small-molecule model heme complexes showed that this spectroscopic signature occurs in bis-imidazole ferrihemes in which the planes of the imidazole ligands are nearly perpendicular, deltaphi = 57-90 degrees. In the last decade protein crystallographic studies have revealed the atomic structures of a number of examples of bis-histidine heme proteins. A frequent characteristic of these large g(max) ferrihemes in membrane-bound proteins is the occurrence of the heme within a four-helix bundle with a left-handed twist. The histidine ligands occur at the same level on two diametrically opposed helices of the bundle. These ligands have the same side-chain conformation and ligate heme iron on the bundle axis, resulting in a quasi-twofold symmetric structure. The two non-ligand-bearing helices also obey this symmetry, and have a conserved small residue, usually glycine, where the edge of the heme ring makes contact with the helix backbones. In many cases this small residue is preceded by a threonine or serine residue whose side-chain hydroxyl oxygen acts as a hydrogen-bond acceptor from the N(delta1) atom of the heme-ligating histidine. The deltaphi angle is thus determined by the common histidine side-chain conformation and the crossing angle of the ligand-bearing helices, in some cases constrained by hydrogen bonds to the serine/threonine residues on the non-ligand-bearing helices.
Product Description
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidine
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Histidine, European Pharmacopoeia (EP) Reference Standard