Merck
  • [Comparative study of the rheological properties of artificial saliva types based on hydroxy-propyl-methyl-cellulose (methocel) and carboxy-methyl-cellulose (CMC)].
[Comparative study of the rheological properties of artificial saliva types based on hydroxy-propyl-methyl-cellulose (methocel) and carboxy-methyl-cellulose (CMC)].
Fogorvosi szemle (1992-02-01)
E Olveti, G Nagy, G Keszthelyi
ABSTRACT
Apparent viscosity at different shear rates was measured for two types of saliva substitutes based on methocel and CMC, and was compared with that of human whole saliva. Results indicate, that fresh mixed human saliva is viscoelastic, exhibiting pseudoplasticity. Methocel containing artificial saliva and human whole saliva are similar in their rheological properties, while the preparation based on CMC is almost a Newtonian liquid. From the results it can be concluded that the saliva substitute containing 0.5% of methocel appears to be a better substitute for natural saliva than of containing CMC as far as rheological properties are concerned.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture