Merck
Rapid methods for deoxynivalenol and other trichothecenes.
Toxicology letters (2004-09-03)
Elisabeth Schneider, Valeriu Curtui, Caroline Seidler, Richard Dietrich, Ewald Usleber, Erwin Märtlbauer
ABSTRACT
Method development for deoxynivalenol (DON) and other trichothecenes in recent years was driven by the analytical necessities arising from its widespread (and increasing) occurrence in foods and feeds. This has resulted in the establishment of guideline levels for animal feed, tolerable daily intake (TDI) levels for humans, and most importantly, in the prospect of low-tolerance levels for these toxins in foods in the near future. In order to ensure reliable determination of the toxin content at the tolerance levels, routine analytical methods must have detection limits of less than the tolerance level. This paper intends to give an overview of current analytical developments of rapid testing for deoxynivalenol and other trichothecene mycotoxins, with a special focus on antibody-based techniques. This includes high-throughput instrumental analysis for the laboratory environment, as well as rapid visual tests for on-site testing. The applicability of rapid tests within an integrated detection system for mycotoxins in foods is discussed.
Sigma-Aldrich
Deoxynivalenol
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard