Merck
  • A decrease in spatially resolved near-infrared spectroscopy-determined frontal lobe tissue oxygenation by phenylephrine reflects reduced skin blood flow.
A decrease in spatially resolved near-infrared spectroscopy-determined frontal lobe tissue oxygenation by phenylephrine reflects reduced skin blood flow.
Anesthesia and analgesia (2014-03-22)
Shigehiko Ogoh, Kohei Sato, Kazunobu Okazaki, Tadayoshi Miyamoto, Frederik Secher, Henrik Sørensen, Peter Rasmussen, Niels H Secher
ABSTRACT
Spatially resolved near-infrared spectroscopy-determined frontal lobe tissue oxygenation (ScO2) is reduced with administration of phenylephrine, while cerebral blood flow may remain unaffected. We hypothesized that extracranial vasoconstriction explains the effect of phenylephrine on ScO2. We measured ScO2 and internal and external carotid as well as vertebral artery blood flow in 7 volunteers (25 [SD 4] years) by duplex ultrasonography during IV infusion of phenylephrine, together with middle cerebral artery mean blood velocity, forehead skin blood flow, and mean arterial blood pressure. During phenylephrine infusion, mean arterial blood pressure increased, while ScO2 decreased by -19% ± 3% (mean ± SE; P = 0.0005). External carotid artery (-27.5% ± 3.0%) and skin blood flow (-25.4% ± 7.8%) decreased in response to phenylephrine administration, and there was a relationship between ScO2 and forehead skin blood flow (Pearson r = 0.55, P = 0.042, 95% confidence interval [CI], = 0.025-0.84; Spearman r = 0.81, P < 0.001, 95% CI, 0.49-0.94) and external carotid artery conductance (Pearson r = 0.62, P = 0.019, 95% CI, 0.13 to 0.86; Spearman r = 0.64, P = 0.012, 95% CI, 0.17-0.88). These findings suggest that a phenylephrine-induced decrease in ScO2, as determined by INVOS-4100 near-infrared spectroscopy, reflects vasoconstriction in the extracranial vasculature rather than a decrease in cerebral oxygenation.
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Supelco
R-(-)-Phenylephrine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Phenylephrine, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard