MilliporeSigma
  • Mood and physical symptoms improve in women with severe cyclical changes by taking an oral contraceptive containing 250-mcg norgestimate and 35-mcg ethinyl estradiol.
Mood and physical symptoms improve in women with severe cyclical changes by taking an oral contraceptive containing 250-mcg norgestimate and 35-mcg ethinyl estradiol.
Contraception (2012-11-06)
Sigrid Nyberg
ABSTRACT
The purpose of this study was to investigate how women without and with different severity of premenstrual symptoms react to treatment with a combined oral contraceptive containing 250-mcg norgestimate/35-mcg ethinyl estradiol (EE). Focus was placed on mood and physical symptoms. This open, prospective study evaluated 24 women using norgestimate/EE for three cycles in a 21/7 regimen. Symptoms and bleeding pattern were captured by daily ratings on the Cyclicity Diagnoser scale. Women with severe premenstrual mood symptoms improved in summarized negative mood (p<.001) and summarized positive mood (p<.05), as well as in swelling (p<.05) and effect on daily life (p<.05). Women with no or mild or moderate symptoms did not show any significant improvement or deterioration in any symptom after 3 months of treatment. Norgestimate 250 mcg/EE 35 mcg significantly improved premenstrual summarized negative mood symptoms during 3 treatment months compared to pretreatment in women with severe premenstrual symptoms, together with improvement in positive symptoms, swelling and effect on daily life.
Product Description
Norgestrel, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
USP
Norgestrel, United States Pharmacopeia (USP) Reference Standard