Merck
  • Right upper quadrant pain and raised alkaline phosphatase is not always a hepatobiliary problem.
Right upper quadrant pain and raised alkaline phosphatase is not always a hepatobiliary problem.
Annals of the Royal College of Surgeons of England (2014-01-15)
G Cheyne, F Runau, D M Lloyd
ABSTRACT
Right upper quadrant pain is a common presenting complaint to the general and hepatobiliary surgical team. Differential diagnoses include gallstones, cholecystitis, liver and pancreatic pathology. A 64-year-old man presented to our general surgical unit with right upper quadrant pain and deranged liver function tests. He underwent ultrasonography several times as well as magnetic resonance cholangiopancreatography (MRCP) in pursuit of hepatobiliary pathology. However, it was the identification of an empyema on MRCP that led to computed tomography of the thorax and the eventual discovery of the cause of the pain: a paraspinal abscess causing T10/T11 discitis. Right upper quadrant pain and deranged liver function tests justify hepatobiliary investigation. Nevertheless, after several negative tests, the differential diagnoses should be broadened and referred pain considered.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host