Merck
  • Learning curve in the application of a hydrogel spacer to protect the rectal wall during radiotherapy of localized prostate cancer.
Learning curve in the application of a hydrogel spacer to protect the rectal wall during radiotherapy of localized prostate cancer.
Urology (2013-10-01)
Michael Pinkawa, Jens Klotz, Victoria Djukic, Carolin Schubert, Nuria Escobar-Corral, Mariana Caffaro, Marc D Piroth, Richard Holy, Michael J Eble
ABSTRACT
To evaluate the effect of increasing experience on hydrogel dimensions, rectal dose, and acute toxicity, and to discuss important technical issues gained from this experience. Sixty-four consecutive patients with prostate cancer were included in this analysis (G1/G2 corresponding to first/second 32 patients) after injection of 10 mL spacer gel. All patients were treated with a 5-field intensity-modulated radiotherapy technique to 76-78 Gy. Treatment toxicity was evaluated with a validated quality of life questionnaire (expanded prostate cancer index composite) before and after radiotherapy. Rectum volume could be entirely excluded from the planning target volume in 31% in G1 vs 56% in G2 (P = .04). Increasing symmetry was detected comparing the first 15 patients to the subsequent rest, with mean differences between right and left of 0.6 cm vs 0.3 cm at the midgland (P = .03). Mean distance between prostate and anterior rectal wall increased from 0.8 cm/1.1 cm/0.8 cm (G1) at the base/middle/apex to 1.3 cm/1.5 cm/1.2 cm (G2), respectively, so that the dose to the rectum decreased significantly (6% vs 2% of the volume inside the 70 Gy isodose; P <.01). Bowel function and bother score changes were smaller comparing baseline with last day of radiotherapy levels (mean 16/18 in G1 vs 9/12 in G2). A learning curve could be demonstrated in our patient population, respecting improved and more symmetrical spacer placement, improved treatment planning, and less treatment-related acute toxicity. Several important technical aspects need to be considered.
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor