Merck
  • Vitamin D3 effects on lipids differ in statin and non-statin-treated humans: superiority of free 25-OH D levels in detecting relationships.
Vitamin D3 effects on lipids differ in statin and non-statin-treated humans: superiority of free 25-OH D levels in detecting relationships.
The Journal of clinical endocrinology and metabolism (2013-09-14)
Lynn Kane, Kelly Moore, Dieter Lütjohann, Daniel Bikle, Janice B Schwartz
ABSTRACT
Inverse associations between 25-OH vitamin D levels and cardiovascular morbidity and mortality have been reported. Our goals were to 1) investigate effects of correcting inadequate D status on lipids, 2) determine whether free 25-OH D is better correlated with lipids than total 25-OH D. A randomized, double-blind placebo-controlled trial was performed. Participants resided in the general community. Adults with inadequate D status were randomized to D3: 14 men, 12 women, age 60 ± 8 years (mean ± SD) or placebo: 12 men, 11 women: 59 ±12 years. Responses to 12-week oral vitamin D3 titrated (1000-3000 IU/d) to achieve 25-OH D levels ≥25 ng/mL were compared to placebo. Measurements were 25-OH D (tandem mass spectometry), free 25-OH D (direct immunoassay), lipids (directly measured triglyceride, cholesterol, and subfractions; plant sterols and cholesterol synthesis precursors), and safety labs before and after 6 and 12 weeks D3 or placebo. Data were analyzed by repeated measures ANOVA and linear regression. Vitamin D3 was titrated to 1000 IU/d in 15/26 (58%), to 2000 IU/d in 10, and 3000 IU/d in one patient. D3 had no effect on cholesterol or cholesterol subfractions except for trends for decreases in atorvastatin-treated patients (cholesterol, P = .08; low-density lipoprotein [LDL] cholesterol, P = .05). Decreased campesterol concentrations (P = .05) were seen with D3 but not placebo in statin-treated patients. Relationships between total 25-OH D and lipids were not detected, but inverse linear relationships were detected between free 25-OH D and triglycerides (P = .03 for all participants [n = 49], P = .03 in all statin-treated [n = 19], and P = .0009 in atorvastatin-treated [n = 11]), and between free 25-OH D and LDL cholesterol (P = .08 overall, P = .02 in all statin-treated, and P = .03 for atorvastatin-treated), and total cholesterol (P = .09 overall; P = .04 for all statin-treated, and P = .05 for atorvastatin-treated). Vitamin D lipid-lowering effects appear limited to statin-treated patients and are likely due to decreased cholesterol absorption. Relationships between lipids and D metabolites were only detected when free 25-OH D was measured, suggesting the superiority of determining free 25-OH D levels compared to total 25-OH vitamin D levels when analyzing biologic responses.
Product Description
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material