Merck
  • Mini-review: Specificity and expression of CIITA, the master regulator of MHC class II genes.
Mini-review: Specificity and expression of CIITA, the master regulator of MHC class II genes.
European journal of immunology (2004-05-27)
Salomé LeibundGut-Landmann, Jean-Marc Waldburger, Michal Krawczyk, Luc A Otten, Tobias Suter, Adriano Fontana, Hans Acha-Orbea, Walter Reith
ABSTRACT
The class II transactivator (CIITA) has been referred to as the "master control factor" for the expression of MHC class II (MHCII) genes. As our knowledge on the specificity and function of CIITA grows, it is becoming increasingly evident that this sobriquet is entirely justified. First, despite extensive investigations, the major target genes of CIITA remain those implicated in the presentation of antigenic peptides by MHCII molecules. Although other putative target genes have been reported, the contribution of CIITA to their expression remains indirect, controversial or comparatively minor relative to its decisive role as a regulator of MHCII and related genes. Second, the most important parameter dictating MHCII expression is by far the expression pattern of the gene encoding CIITA (MHC2TA). The vast majority of signals that activate or repress MHCII expression under physiological and pathological situations converge on one or more of the three alternative promoters that drive transcription of the MHC2TA gene. In short, with respect to its specificity and its exquisitely controlled pattern of expression, CIITA is by a long stretch the single most important transcription factor for the regulation of genes required for MHCII-restricted antigen-presentation.
Sigma-Aldrich
Anti-CIITA antibody produced in rabbit, affinity isolated antibody