Merck
  • Use of high-resolution proton nuclear magnetic resonance spectroscopy for rapid multi-component analysis of urine.
Use of high-resolution proton nuclear magnetic resonance spectroscopy for rapid multi-component analysis of urine.
Clinical chemistry (1984-03-01)
J R Bales, D P Higham, I Howe, J K Nicholson, P J Sadler
ABSTRACT
Numerous low-Mr metabolites--including creatinine, citrate, hippurate, glucose, ketone bodies, and various amino acids--have been identified in 400- and 500-MHz proton nuclear magnetic resonance (1H NMR) spectra of intact human urine. The presence of many of these was related to the specific condition of the donors: humans in different physiological states (resting, fasting, or post-exercise) and pathological conditions (e.g., diabetes mellitus, cadmium-induced renal dysfunction). We have also monitored the metabolism of simple nonendogenous compounds (methanol and ethanol) and of acetaminophen. The pH-dependencies of the NMR chemical shifts of some urine components are reported. Our studies show that high-resolution 1H NMR spectroscopy provides a fast, simple method for "fingerprint" identification of urinary compounds. In some cases, analytes can be quantified by standard additions or by comparing integrated peak areas for the metabolites with those for creatinine. Determinations of creatinine by 1H NMR spectroscopy compared well with those by an independent chemical assay based on the Jaffé reaction.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
L-(+)-Lactic acid solution, 30% in H2O (by weight)
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Supelco
Acetone, analytical standard
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)