Merck
  • Suppression of reperfusion-induced arrhythmias with combined administration of 5-HT2 and thromboxane A2 antagonists.
Suppression of reperfusion-induced arrhythmias with combined administration of 5-HT2 and thromboxane A2 antagonists.
British journal of pharmacology (1996-03-01)
L A Shaw, S J Coker
ABSTRACT
1. The effects of the 5-HT2 antagonist, ICI 170,809 and the thromboxane A2 antagonist, ICI 192,605, given alone and in combination (n = 12 per group), were examined in anaesthetized rats. Haemodynamics and arrhythmias induced by permanent coronary artery occlusion or by reperfusion after 5 min of ischaemia were monitored. 2. In a study on reperfusion-induced arrhythmias, the only significant effect of ICI 170,809 (1 mg kg-1, i.v.) was a reduction in the number of ventricular premature beats (VPBs). ICI 192,605 (1 mg kg-1 min-1, i.v.) did not alter reperfusion-induced arrhythmias. However, in combination, when compared with controls, these drugs caused significant reductions in the incidence of ventricular tachycardia (VT), 100% to 58%; ventricular fibrillation (VF), 92% to 33%; and the mortality due to sustained VF, 67% to 17%. There was also a significant reduction in the number of VPBs following reperfusion. 3. In a second study with lower doses of drugs, ICI 170,809 (0.3 mg kg-1) and ICI 192,605 (0.3 mg kg-1 min-1) had no significant effects on reperfusion-induced arrhythmias either alone or in combination. 4. A third study examined the effects of the higher doses of the drugs on ischaemia-induced arrhythmias. Neither drug alone, nor in combination, altered the incidence of ischaemia-induced VT, VF, the mortality, or the number of VPBs. 5. These results indicate that, in contrast to the administration of either drug alone, combined administration of a 5-HT2 antagonist and a thromboxane A2 antagonist caused marked suppression of reperfusion-induced but not ischaemia-induced arrhythmias.
Sigma-Aldrich
ICI 192605, ≥98% (HPLC)