Merck
Purification and properties of Aspergillus niger beta-glucosidase.
European journal of biochemistry (1992-10-15)
T Watanabe, T Sato, S Yoshioka, T Koshijima, M Kuwahara
ABSTRACT
Beta-glucosidase was purified from a crude cellulase preparation from Aspergillus niger by affinity chromatography on a methacrylamide-N-methylene-bis-methacrylamide copolymer bearing cellobiamine. The purified enzyme was a dimer with an isoelectric point of 4.0. The molecular mass of the enzyme was estimated to be 240 kDa by gel-permeation chromatography. The enzyme hydrolyzed specifically beta-glucosidic bonds and catalyzed transglucosylation of the beta-glucosyl group of cellobiose to yield 4-O-beta-gentiobiosylglucose in the presence of organic solvents or under neutral conditions.
Sigma-Aldrich
Glucosidase from Aspergillus niger, powder, ≥750 U/g