Merck
  • Identification and spectral characterization of the external aldimine of the O-acetylserine sulfhydrylase reaction.
Identification and spectral characterization of the external aldimine of the O-acetylserine sulfhydrylase reaction.
Biochemistry (1995-09-26)
K D Schnackerz, C H Tai, J W Simmons, T M Jacobson, G S Rao, P F Cook
ABSTRACT
The O-acetylserine sulfhydrylase (OASS) reaction has been studied using a number of spectral probes including UV--visible, fluorescence, circular dichroism, and 31P NMR spectroscopy. The addition of L-cysteine, L-alanine, and glycine to OASS results in a shift in lambda max of 412 nm for the internal Schiff base to 418 nm resulting from the formation of the external Schiff base. The addition of L-serine or O-methyl-D,L-serine gives decreases of the absorbance of unliganded enzyme at 412 nm of about 50% and 20%, respectively, concomitant with an increase in the absorbance at 320 nm and a shift in the lambda max of the remaining visible absorbance to 418 nm. The spectral shifts observed in the presence of L-serine are suggestive of establishing an equilibrium between different forms of external Schiff base. The concentration dependence of the changes at 440 (L-cysteine) and 320 nm (L-serine) provides an estimate of the dissociation constant for the external aldimine. The pH dependence of the dissociation constant suggests the alpha-amine of the amino acid must be unprotonated for nucleophilic attack at C4' of PLP, and an enzyme side chain must be unprotonated to hydrogen-bond the thiol or hydroxyl side chain of the amino acid. When L-cysteine is the amino acid, the thiol side chain must be protonated to hydrogen-bond to the unprotonated enzyme side chain. The 31P NMR chemical shift is increased from 5.2 ppm for unliganded enzyme to 5.3 ppm in the presence of L-cysteine, signaling a tighter interaction at the 5'-phosphate upon formation of the external Schiff base.(ABSTRACT TRUNCATED AT 250 WORDS)