Merck
  • Incorporation of deuterium-labeled trans- and cis-13-octadecenoic acids in human plasma lipids.
Incorporation of deuterium-labeled trans- and cis-13-octadecenoic acids in human plasma lipids.
Journal of lipid research (1983-01-01)
E A Emken, R O Adlof, W K Rohwedder, R M Gulley
ABSTRACT
The absorption and distribution of deuterated trans- and cis-13-octadecenoic acid (13t-18:1 and 13c-18:1) in plasma lipids were compared to deuterated cis-9-octadecenoic acid (9c-18:1) in two young adult male subjects. A mixture of triglycerides was fed in a multiple-labeled experiment where each triglyceride contained a fatty acid labeled with a different number of deuterium atoms. Analysis of human plasma lipids by mass spectroscopy allowed the distribution of the two 13-octadecenoic acid isomers to be directly compared to cis-9-octadecenoic acid. Plasma lipids selectively excluded both the 13t-18:1 and 13c-18:1 isomers relative to 9c-18:1 in all neutral and phospholipid fractions. Discrimination against incorporation of the 13t-18:1 isomer into plasma cholesteryl ester and 2-acyl phosphatidylcholine was nearly absolute. The 1-acyl phosphatidylcholine fraction exhibited a large positive selectivity for the 13t-18:1 isomer. Differences in the relative distribution of the trans and cis 13-18:1 isomers vs. 9c-18:1 in the various lipoprotein lipid classes were found. Analysis of the chylomicron triglyceride component of the plasma lipids indicated all three fatty acids were equally well absorbed.