Merck
  • Loss of tritium from coprostanone derived from [1,2(n)-3H]cholesterol or [7(n)-3H]cholesterol.
Loss of tritium from coprostanone derived from [1,2(n)-3H]cholesterol or [7(n)-3H]cholesterol.
Journal of lipid research (1980-05-01)
G E Mott, E M Jackson
ABSTRACT
After oral administration of a mixture of [1,2(n)-3H]cholesterol and [4-14C]cholesterol to a baboon, fecal coprostanone had a 46% lower 3H/14C ratio than the dose administered. Loss of 3H by enolization of the 3-ketone could account for the decrease in 3H/14C. If [7(n)-3H]cholesterol was administered instead of [1,2(n)-3H]cholesterol a 23% loss of 3H from coprostanone was found. Procedures requiring measurement of 3H-coprostanone derived from [1,2(n)-3H]- or [7(n)-3H]cholesterol could be seriously in error unless an appropriate correction for loss of 3H is made.
Sigma-Aldrich
5α-Cholestan-3-one, crystalline