Merck
  • Characterization of a neutral aminoacyl-peptide hydrolase from Naegleria fowleri.
Characterization of a neutral aminoacyl-peptide hydrolase from Naegleria fowleri.
The Journal of protozoology (1987-05-01)
F Marciano-Cabral, S Stanitski, V Radhakrishna, S G Bradley
ABSTRACT
An intracellular alpha-aminoacyl-peptide hydrolase (EC 3.4.11.-) from Naegleria fowleri nN68 (ATCC 30894) has been characterized. The enzyme preparation hydrolyzed phenylalanyl-, tyrosyl-, leucyl-, arginyl-, alanyl-, tryptophanyl-, histidyl-, methionyl-, and lysyl-naphthylamide but not benzoylleucyl-, leucylglycyl-, glycylprolylleucyl-, glycyl-, threonyl-, aspartyl-, or glutamyl-naphthylamide. The aminopeptidase activity was inhibited by the cysteine-protease inhibitors--hydroxymercuribenzoate, chloromercurisulfate, and iodoacetate--by the aminopeptidase inhibitors--bestatin and trans-epoxysuccinyl-leucyl-agmatine--by an inhibitor of soluble alanyl aminopeptidase EC 3.4.11.14, puromycin, and by the metalloprotease inhibitor, o-phenanthroline. The exopeptidase activity was not inhibited by the chelator, ethylenediaminetetraacetate, or the serine-protease inhibitor, phenylmethylsulfonylfluoride. The pH optimum of the exopeptidase was between 7.0 and 8.0. Enzyme activity was stable at 55 degrees C for 30 min, but all activity was lost after 15 min at 80 degrees C. Enzyme activity was inhibited by 100 microM HgCl2 and CdCl2 but not by 1 mM CoCl2, CuCl2, MnCl2, NiCl2, FeCl3, or ZnCl2. Enzyme activity was inhibited by 0.1% sodium dodecyl sulfate but not by 0.2% Brij 35, Tween 20, Tween 80, or Triton X-100.
Sigma-Aldrich
L-Leucine β-naphthylamide