Merck
  • Effect of β-(3,4-dihydroxyphenyl)lactic acid on oxidative stress stimulated by high glucose levels in human peritoneal mesothelial cells.
Effect of β-(3,4-dihydroxyphenyl)lactic acid on oxidative stress stimulated by high glucose levels in human peritoneal mesothelial cells.
The Journal of international medical research (2012-08-22)
H Zhang, J-W Wang, Y Xu, K Zhang, B Yi, J Sun, Y Liu, X-M Zhang, J-S Liu
ABSTRACT
To investigate the effects of β-(3,4-dihydroxyphenyl)lactic acid on oxidative stress stimulated by high glucose levels in human peritoneal mesothelial cells (HPMCs) in vitro. HPMCs were incubated with 100 mol/l glucose followed by 0.625-20 mg/ml β-(3,4-dihydroxyphenyl)lactic acid. Reactive oxygen species (ROS) were quantified by flow cytometry. Relative levels of fibronectin-1 (FN1), collagen-I α(1) (COL1A1), endothelin-1 (EDN1) and haem oxygenase-1 (HMOX1) mRNA and protein were quantified by real-time reverse transcription-polymerase chain reaction and Western blotting, respectively. Absolute levels of FN1 and COLIA1 were quantified by enzyme-linked immunosorbent assay. β-(3,4-Dihydroxyphenyl)lactic acid significantly decreased ROS levels, and EDN1 mRNA and protein levels, in dose- and time-dependent manners. HMOX1 mRNA and protein levels were significantly increased by β-(3,4-dihydroxyphenyl)lactic acid in dose-dependent manners. COL1A1 and FN1 mRNA and protein levels were significantly decreased by β-(3,4-dihydroxyphenyl)lactic acid in dose- and time-dependent manners. β-(3,4-Dihydroxyphenyl)lactic acid inhibited oxidative stress and reversed increases in FN1 and COLIA1 induced by high glucose levels in HPMCs. This may contribute to a protective role in peritoneal fibrosis.
Sigma-Aldrich
Danshensu sodium salt, ≥98% (HPLC)