Merck
  • Physico-chemical properties of Salmonella typhi Vi polysaccharide-diphtheria toxoid conjugate vaccines affect immunogenicity.
Physico-chemical properties of Salmonella typhi Vi polysaccharide-diphtheria toxoid conjugate vaccines affect immunogenicity.
Vaccine (2011-08-17)
So Jung An, Yeon Kyung Yoon, Sudeep Kothari, Neha Kothari, Jeong Ah Kim, Eugene Lee, Deok Ryun Kim, Tai Hyun Park, Greg W Smith, Rodney Carbis
ABSTRACT
In this study it was demonstrated that the immunogenicity of Vi polysaccharide-diphtheria toxoid conjugates was related to the physical and chemical structure of the conjugate. Conjugates were prepared in two steps, firstly binding adipic acid dihydrazide (ADH) spacer molecules to diphtheria toxoid (DT) carrier protein then secondly binding varying amounts of this derivatized DT to a fixed amount of Vi capsular polysaccharide purified from Salmonella enterica Serovar Typhi. As the amount of DT bound to the Vi increased the size of the conjugate increased but also the degree of cross-linking increased. The immunogenicity of the conjugates was tested in mice and measured by ELISA for anti Vi and anti DT IgG responses, and the results revealed a trend that as the amount of DT bound to the Vi increased the anti Vi responses increased. This study establishes a correlation between physico-chemical characteristics of the conjugate and the magnitude of the anti Vi and anti DT responses.
Sigma-Aldrich
Adipic acid dihydrazide, ≥98% (titration)