Merck
  • Potential compounds for monitoring method performance in the determination of selected organochlorine pesticides and polychlorinated biphenyls in human serum.
Potential compounds for monitoring method performance in the determination of selected organochlorine pesticides and polychlorinated biphenyls in human serum.
Journal of analytical toxicology (1990-05-01)
V W Burse, P C McClure, M P Korver, S L Head, L L Needham
ABSTRACT
Four compounds--2,2', 3,4'-tetrachlorobiphenyl, decachlorodiphenylether, 1,1-dichloro-2,2-bis(p-ethylphenyl)ethane, and dichlorobenzophenone--are recommended for monitoring the within-sample behavior of an analytical method that quantifies chlorinated pesticides and polychlorinated biphenyls (such as Aroclor 1260) in serum using packed column gas chromatography with electron capture detection. Percent recoveries of these surrogates averaged greater than 80%, except with dichlorobenzophenone, which had an average recovery of greater than 70%.
Sigma-Aldrich
4,4′-Dichlorobenzophenone, 99%