MilliporeSigma
  • Assay of glyceryl trinitrate, isosorbide dinitrate, and their metabolites in plasma by large-bore capillary column gas-liquid chromatography.
Assay of glyceryl trinitrate, isosorbide dinitrate, and their metabolites in plasma by large-bore capillary column gas-liquid chromatography.
Biopharmaceutics & drug disposition (1990-11-01)
B P Booth, B M Bennett, J F Brien, D A Elliott, G S Marks, J L McCans, K Nakatsu
ABSTRACT
Two large-bore capillary columns, one with dimethyl polysiloxane (HP-1) as the stationary phase and the other with phenyl (50 per cent) methyl (50 per cent) polysiloxane (DB-17), were used to develop gas-liquid chromatographic (GLC) assays for measuring isosorbide dinitrate (ISDN), glyceryl trinitrate (GTN), and their metabolites. ISDN, isosorbide-2-mononitrate (2-ISMN), and isosorbide-5-mononitrate (5-ISMN) in plasma, ranging in concentration from 1 to 300 nM, and GTN, glyceryl-1,2-dinitrate (1,2-GDN), and glyceryl-1,3-dinitrate (1,3-GDN), ranging in concentration from 3 to 60 nM in plasma, were analysed on both columns. GLC analysis yielded baseline resolution of the analytes. The method using the dimethyl polysiloxane column gave a lower limit of detectability for GTN of 0.75 nM (signal/noise (s/n) = 2), and the procedure using the phenyl-methyl column provided a lower limit of detectability for ISDN of 81 pM (s/n = 2). The large-bore column GLC procedures exhibited shorter retention times for both ISDN and GTN than those previously reported for capillary-column assays. The chromatographic resolution of analytes and column efficiency of the large-bore capillary columns were comparable to the results previously found using capillary-column GC. The assays for ISDN and GTN have been shown to be appropriate for pharmacokinetic studies in volunteers and patients. We determined that the HP-1 column is appropriate for the analysis of GTN and metabolites, and the DB-17 column is suitable for analysis of ISDN and its metabolites. We conclude that the use of large-bore capillary columns provides rapid and reliable GLC assays for organic nitrates.
Product Description
Isosorbide 2-nitrate, European Pharmacopoeia (EP) Reference Standard