Merck
  • Folded structures of L-leucylglycine oligopeptides and their aggregational behavior in aqueous solution: Raman scattering spectra and proton NMR spin-lattice relaxation studies.
Folded structures of L-leucylglycine oligopeptides and their aggregational behavior in aqueous solution: Raman scattering spectra and proton NMR spin-lattice relaxation studies.
The journal of physical chemistry. B (2008-04-15)
Aki-Hiro Yoshino, Hiro-Fumi Okabayashi, Hide-Hiro Kanbe, Keita Suzuki, Charmian J O'Connor
ABSTRACT
The aggregational behavior of three L-leucylglycine oligopeptides (residue numbers of glycine are 3, 4, and 5) in aqueous solution was investigated by the use of Raman scattering and 1H NMR spin-lattice relaxation methods. The results indicate that their oligopeptides take up a folded structure to form dimeric aggregates above their critical aggregation concentration. The application of one-dimensional aggregate theory to these systems provides the following prediction. Elongation up to 6 glycine residues makes it possible to form dimeric aggregates, but further elongation (up to 7 glycine residues) makes the aggregates very unstable, and up to 8 or 9 glycine residues makes the formation of dimeric aggregates very difficult. The one-dimensional aggregate theory may be used to predict the existence of peptide aggregates through intermolecular hydrogen bonding.