Merck
  • 3-Aminorhodanine and 2-hydrazino-2-imidazo-line hydrazones. Synthesis and antiparasitic pharmacology.
3-Aminorhodanine and 2-hydrazino-2-imidazo-line hydrazones. Synthesis and antiparasitic pharmacology.
Bollettino chimico farmaceutico (1991-04-01)
N E Madadi, P Vanelle, J Maldonado, F Tedlaouti, B Majester, M Gasquet, F Delmas, P Timon-David
ABSTRACT
Two series of hydrazones were prepared by condensation of various aldehydes with 2-aminorhodanine (series 1) and 2-hydrazino-2-imidazoline (series 2). These compounds showed interesting antiparasitic activities against amoeba, trichomonas, leishmania and plasmodium.
Sigma-Aldrich
3-Aminorhodanine, 99%