Merck
  • Purification, characterization, and crystallization of human pyrroline-5-carboxylate reductase.
Purification, characterization, and crystallization of human pyrroline-5-carboxylate reductase.
Protein expression and purification (2006-04-08)
Zhaohui Meng, Zhiyong Lou, Zhe Liu, Dong Hui, Mark Bartlam, Zihe Rao
ABSTRACT
Pyrroline-5-carboxylate reductase (P5CR) catalyzes the reduction of Delta1-pyrroline-5-carboxylate (P5C) to proline with concomitant oxidation of NAD(P)H to NAD(P)(+). The enzymatic cycle between P5C and proline is very important in many physiological and pathological processes. Human P5CR was over-expressed in Escherichia coli and purified to homogeneity by chromatography. Enzymatic assays of the wild-type protein were carried out using 3,4-dehydro-L-proline as substrate and NAD(+) as cofactor. The homopolymer was characterized by cross-linking and size exclusion gel filtration chromatography. Human P5CR was crystallized by the hanging-drop vapor-diffusion method at 37 degrees C. Diffraction data were obtained to a resolution of 2.8A and were suitable for high resolution X-ray structure determination.
Sigma-Aldrich
3,4-Dehydro-DL-proline, 98%