Merck
  • Two-dimensional supramolecular arrangements of enantiomers and racemic modification of 1,1'-binaphthyl-2,2'-dicarboxylic acid.
Two-dimensional supramolecular arrangements of enantiomers and racemic modification of 1,1'-binaphthyl-2,2'-dicarboxylic acid.
Langmuir : the ACS journal of surfaces and colloids (2005-09-21)
Masashi Kunitake, Tetsutaro Hattori, Sotaro Miyano, Kingo Itaya
ABSTRACT
Formation of adlayers of the optically active compound 1,1'-binaphthyl-2, 2'-dicarboxylic acid (BINAC) on iodine-modified Au (111) surfaces in perchloric acid was investigated by in situ scanning tunneling microscopy (STM). Highly ordered arrays formed on the surfaces via simple spontaneous adsorption from a solution of enantiomers or the racemic BINAC, in spite of the fact that BINAC has a three-dimensionally complex stereochemical structure. Adlayers of both enantiomers essentially shared the same enantiomorphous structure. Observed parameters of the rectangular unit cell lattice for arrays of both enantiomers of BINAC were a = 2.3 +/- 0.2 nm and b = 0.7 +/- 0.2 nm. On the other hand, racemic modification formed an entirely different adlayer, which consisted of an alternate alignment of the two enantiomers, with an oblique unit cell lattice with parameters of a = 1.2 +/- 0.2 nm, b = 0.8 +/- 0.1 nm, and 74 +/- 3 degrees . No domain composed of a single enantiomer was observed. The stronger hetero-intermolecular interactions of enantiomer couples led to the formation of an alternate arrangement in the array prepared by racemic modification.
Sigma-Aldrich
Diphenic acid, 97%