Merck
Complementarity in bimolecular photochromism.
Chemical communications (Cambridge, England) (2005-05-19)
David Bailey, Vance E Williams
ABSTRACT
Irradiating 2,3,6,7-tetraphenylanthracene in the presence of 9,10-dimethylanthracene leads to exclusive formation of the cross-dimer. No photochemical reaction is observed when either of these chromophores is irradiated in the absence of the other.
Sigma-Aldrich
9,10-Dimethylanthracene, 99%