Merck
  • Rapid enantioseparation of 1-aminoindan by microchip electrophoresis with linear-imaging UV detection.
Rapid enantioseparation of 1-aminoindan by microchip electrophoresis with linear-imaging UV detection.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2005-01-29)
Fumihiko Kitagawa, Shiori Aizawa, Koji Otsuka
ABSTRACT
Chiral separations of 1-aminoindan (AI) by cyclodextrin electrokinetic chromatography (CDEKC) were investigated on microfluidic quartz chips. By using a microchip electrophoresis (MCE) instrument equipped with a linear-imaging UV detector, the separation process of the enantiomeric compounds was observed. When sulfated beta-cyclodextrin was employed as a chiral selector, the baseline separation of AI could be achieved within 1 min with a high repeatability. The relative standard deviation of the migration time was less than 6%. The fastest separation was achieved in 14 s, utilizing a separation length of only 6.1 mm. These results show that the MCE analysis employing a linear imaging UV detector has a significant potential for fast chiral analysis.
Sigma-Aldrich
1-Aminoindan, 98%
Pricing and availability is not currently available.