Merck
  • Comparison of selective media for coagulase-positive enterotoxigenic Staphylococcus aureus.
Comparison of selective media for coagulase-positive enterotoxigenic Staphylococcus aureus.
Applied and environmental microbiology (1978-06-01)
A Niskanen, M Aalto
PMID677885
ABSTRACT
Different culture media were used to detect enterotoxigenic Staphylococcus aureus strains from peptone salt solution and inoculated minced meat. When recovery, reactions, and counting mistakes were considered, it was concluded that Baird-Parker agar was the best medium.
Product Description
Millipore
Baird-Parker Agar, NutriSelect® Basic, powder, final pH 6.8±0.2 (25 °C), pkg of 500 g, non-sterile
Pricing and availability is not currently available.