Merck
  • Potential estrogenic and antiestrogenic activity of the cyclic siloxane octamethylcyclotetrasiloxane (D4) and the linear siloxane hexamethyldisiloxane (HMDS) in immature rats using the uterotrophic assay.
Potential estrogenic and antiestrogenic activity of the cyclic siloxane octamethylcyclotetrasiloxane (D4) and the linear siloxane hexamethyldisiloxane (HMDS) in immature rats using the uterotrophic assay.
Toxicological sciences : an official journal of the Society of Toxicology (2001-08-18)
J M McKim, P C Wilga, W J Breslin, K P Plotzke, R H Gallavan, R G Meeks
ABSTRACT
The cyclic siloxane octamethylcyclotetrasiloxane (D4) and the linear siloxane hexamethyldisiloxane (HMDS) have numerous industrial and consumer applications and thus have the potential for human exposure. The present study was undertaken to examine potential estrogenic and antiestrogenic activities of D4 and HMDS. To address potential differences in sensitivity between rat strains the study used both Sprague-Dawley (SD) and Fischer 344 (F-344) rats. Estrogenicity of the test compounds was determined by measuring absolute and relative uterine weights in immature rats and by monitoring uterine epithelial cell height. In order to place the data obtained for D4 into perspective relative to strong and weak estrogenic compounds, the response produced by D4 at 0, 10, 50, 100, 250, 500, and 1000 mg/kg/day was compared to responses produced by ethinyl estradiol (EE) (1, 3, 10, or 30 microg/kg/day), diethylstilbestrol dipropionate (DES-DP) (0.5, 1.5, 5, 15 microg/kg/day), and coumestrol (CE) (10, 35, 75, 150 mg/kg/day). Antiestrogenic effects were evaluated by co-administering D4 (500 mg/kg/day) with EE at 1, 3, 10, and 30 microg /kg/day. All compounds were administered in sesame oil at a volume of 5 mL/kg by oral gavage. Beginning on postnatal day 18 (SD) or 21 (F-344) each pup (12 per group) received a single dose of test compound once a day for 4 consecutive days. The pups were euthanized the morning after the last treatment and their uteri removed, weighed, and processed for histological examination. EE and DES-DP produced a significant dose-dependent increase in absolute and relative uterine weights and uterine cell height. The maximum increase in uterine weight following EE exposure was approximately 350% relative to controls in both strains. The weak phytoestrogen CE also produced a dose-related increase in absolute and relative uterine weight and epithelial cell height, but the response occurred over a much higher range of doses. At the highest dose of CE, uterine weight was increased approximately 230% relative to controls. Following exposure to D4, absolute and relative uterine weights and uterine epithelial cell height were statistically significantly increased in both strains of rats at doses above 100 mg/kg/day. In terms of uterine weight, D4 was approximately 0.6 million times less potent than EE or DES-DP in SD pups and 3.8 million times less potent than EE or DES-DP in F-344 pups. The maximal increase in uterine weight, relative to controls, produced by D4 at 1000 mg/kg/day was approximately 160% in SD rats, while the maximum increase produced by D4 in F-344 rats was 86%. D4 co-administered over a wide range of EE doses, resulted in a significant reduction in uterine weight compared to EE alone. HMDS was evaluated in SD rats only. The response produced by HMDS (600 and 1200 mg/kg/day) was compared to EE (3 microg/kg/day). Antiestrogenic effects were evaluated by co-administering HMDS (1200 mg/kg/day) with EE at 3 microg/kg/day. HMDS had no measurable effect on uterine weight under the experimental conditions described here. However, HMDS coadministered with EE did produce a small, but statistically significant reduction in uterine weight compared to EE alone. In conclusion, D4 showed weak estrogenic and antiestrogenic activity that was several orders of magnitude less potent than EE, and many times less potent than the weak phytoestrogen CE.
Sigma-Aldrich
Octamethylcyclotetrasiloxane, 98%