Merck
  • Fluorogenic and fluorescent labeling reagents with a benzofurazan skeleton.
Fluorogenic and fluorescent labeling reagents with a benzofurazan skeleton.
Biomedical chromatography : BMC (2001-08-17)
S Uchiyama, T Santa, N Okiyama, T Fukushima, K Imai
ABSTRACT
Fluorogenic and fluorescent labeling reagents having a benzofurazan (2,1,3-benzoxadiazole) skeleton such as 4-fluoro-7-nitro-2,1,3-benzoxadiazole (NBD-F), 4-N,N-dimethylaminosulfonyl-7-fluoro-2,1,3-benzoxadiazole (DBD-F), 4-aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole (ABD-F), ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate (SBD-F), 4-hydrazino-7-nitro-2,1,3-benzoxadiazole (NBD-H), 4-N,N-dimethylaminosulfonyl-7-hydrazino-2,1,3-benzoxadiazole (DBD-H), 4-nitro-7-N-piperazino-2,1,3-benzoxadiazole (NBD-PZ), 4-N,N-dimethylaminosulfonyl-7-N-piperazino-2,1,3-benzoxadiazole (DBD-PZ), 4-(N-chloroformylmethyl-N-methyl)amino-7-N,N-dimethylaminosulfonyl-2,1,3-benzoxadiazole (DBD-COCl) and 7-N,N-dimethylaminosulfonyl-4-(2,1,3-benzoxadiazolyl) isothiocyanate (DBD-NCS) are reviewed in terms of synthetic method, reactivity, fluorescence characteristics, sensitivity and application to analytes.
Sigma-Aldrich
Benzofurazan, 97%