MilliporeSigma
  • Molecular imprinting of biotin derivatives and its application to competitive binding assay using nonisotopic labeled ligands.
Molecular imprinting of biotin derivatives and its application to competitive binding assay using nonisotopic labeled ligands.
Analytical chemistry (2000-06-17)
T Takeuchi, A Dobashi, K Kimura
ABSTRACT
Synthetic biotin-binding polymers were prepared by molecular imprinting. Methacrylic acid (MAA) was copolymerized with ethylene glycol dimethacrylate in the presence of biotin methyl ester (B-Me) in chloroform. Hydrogen-bonding-based complexation of B-Me with MAA generates the binding sites complementary to B-Me after extracting B-Me from the resulting copolymers. Data from NMR titration suggest a one-to-one prepolymerization complex formation of B-Me with MAA in chloroform. A possible complex structure was estimated by docking of the most stable conformers by intermolecular Monte Carlo conformational search under the assumption of a one-to-one association. The selectivity of the imprinted polymers was investigated and an imprinted polymer-based competitive binding assay for B-Me was demonstrated using biotin p-nitrophenyl ester as a nonisotopic-labeled ligand.