Merck
  • The neurotoxic effects of methylenedioxymethamphetamine (MDMA) and its metabolites on rat brain spheroids in culture.
The neurotoxic effects of methylenedioxymethamphetamine (MDMA) and its metabolites on rat brain spheroids in culture.
Cell biology and toxicology (1999-12-02)
T M Walker, J E Davenport-Jones, R M Fox, C K Atterwill
ABSTRACT
Rat whole-brain spheroids were used to assess the intrinsic neurotoxicity of methylenedioxy-methamphetamine (MDMA, Ecstasy) and two of its metabolites, dihydroxymethamphetamine (DHMA) and 6-hydroxy-MDMA (6-OH MDMA). Exposure of brain spheroids to MDMA or the metabolite 6-OH MDMA (up to 500 micromol/L) for 5 days in culture did not alter intracellular levels of glutathione (GSH), glial fibrillary acidic protein (GFAP) or serotonin (5-HT). In contrast, exposure to the metabolite DHMA, which can deplete intracellular thiols, significantly increased GSH levels (up to 170% of control) following exposure to 50 and 100 micromol/L DHMA. There was also a significant reduction in the levels of glial fibrillary acidic protein (GFAP) and GSH by DHMA at the highest concentration tested (500 micromol/L) but there was no effect on 5HT. This may constitute a sublethal neurotoxic compensatory response to DHMA in an attempt to replenish depleted intraneural GSH levels following metabolite exposure. Rat whole-brain spheroids may thus be a useful in vitro model to delineate mechanisms and effects of this class of neurotoxin.