Merck
  • Effects of divalent metal ions on the activity and conformation of native and 3-fluorotyrosine-PvuII endonucleases.
Effects of divalent metal ions on the activity and conformation of native and 3-fluorotyrosine-PvuII endonucleases.
European journal of biochemistry (1999-04-02)
C M Dupureur, L M Hallman
ABSTRACT
The activities of restriction enzymes are important examples of Mg(II)-dependent hydrolysis of DNA. While a number of crystallographic studies of enzyme-DNA complexes have also involved metal ions, there have been no solution studies exploring the relationship between enzyme conformation and metal-ion binding in restriction enzymes. Using PvuII restriction endonuclease as a model system, we have successfully developed biosynthetic fluorination and NMR spectroscopy as a solution probe of restriction-enzyme conformation. The utility of this method is demonstrated with a study of metal-ion binding by PvuII endonuclease. Replacement of 74% (+/- 10%) of the Tyr residues in PvuII endonuclease by 3-fluorotyrosine produces an enzyme with Mg(II)-supported specific activity and sequence specificity that is indistinguishable from that of the native enzyme. Mn(II) supports residual activity of both the native and fluorinated enzymes; Ca(II) does not support activity in either enzyme, a result consistent with previous studies. 1H- and 19F-NMR spectroscopic studies reveal that while Mg(II) does not alter the enzyme conformation, the paramagnetic Mn(II) produces both short-range spectral broadening and longer range changes in chemical shift. Most interestingly, Ca(II) binding perturbs a larger number of different resonances than Mn(II). Coupled with earlier mutagenesis studies that place Ca(II) in the active site [Nastri, H. G., Evans, P.D., Walker, I.H. & Riggs, P.D. (1997) J. Biol. Chem. 272, 25761-25767], these data suggest that the enzyme makes conformational adjustments to accommodate the distinct geometric preferences of Ca(II) and may play a role in the inability of this metal ion to support activity in restriction enzymes.
Sigma-Aldrich
m-Fluoro-DL-tyrosine