Merck
  • cis-1,4-diaminocyclohexane-Pt(II) and -(IV) adducts with DNA bases and nucleosides.
cis-1,4-diaminocyclohexane-Pt(II) and -(IV) adducts with DNA bases and nucleosides.
Journal of inorganic biochemistry (2003-09-19)
Mohammad S Ali, Abdul R Khokhar
ABSTRACT
A series of new platinum(II) and platinum(IV) adducts of type [P(II)(cis-1,4-DACH)LCl]NO(3,) where cis-1,4-DACH=cis-1,4-diaminocyclohexane, and L=9-ethylguanine, 1-methylcytosine, adenine, adenosine, cytosine, cytidine, guanine, and [Pt(IV)(cis-1,4-DACH)Ltrans-(X)(2)Cl]NO(3), (where Y=hydroxo or acetato), were synthesized and characterized by elemental analysis, infrared spectroscopy, and 1H and 195Pt nuclear magnetic resonance spectroscopy.
Sigma-Aldrich
trans-1,4-Diaminocyclohexane, ≥98.0% (GC)