Merck
  • Structure-function relationship in glycosylated alpha-chymotrypsin as probed by IMAC and IMACE.
Structure-function relationship in glycosylated alpha-chymotrypsin as probed by IMAC and IMACE.
Biochimica et biophysica acta (1999-08-14)
K Y Jiang, O Pitiot, M Anissimova, H Adenier, M A Vijayalakshmi
ABSTRACT
Chemical glycosylation of bovine alpha-chymotrypsin, by a glucosamine adduct on the carboxyl group, results in the modification of its catalytic activity. The structural alterations of alpha-chymotrypsin resulting from its glycosylation are studied by immobilized metal-ion affinity chromatography (IMAC) and immobilized metal-ion affinity capillary electrophoresis (IMACE). The chemical glycosylation of alpha-chymotrypsin generates two distinct subpopulations of the protein: one which totally loses the initial affinity for IDA-Cu(II) and another which exhibits an increased affinity for the metal chelate ligand.
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
N-Benzoyl-L-tyrosine ethyl ester