Merck
  • [Beneficial effects of immunostimulant and antiviral therapy of ophthalmic herpes zoster].
[Beneficial effects of immunostimulant and antiviral therapy of ophthalmic herpes zoster].
Romanian journal of virology (1993-01-01)
V Topciu, G Chercotă, R Mihăilescu, A Nadolnic
ABSTRACT
A group of 16 patients with ophthalmic zona zoster received antiviral and immunostimulative treatment with Romanian specific products. Results were spectacular. The problem of the identity of the varicella and zoster viruses is discussed.
Sigma-Aldrich
Moroxydine hydrochloride, 99%