Merck
  • Retinoic acid receptor beta,gamma-selective ligands: synthesis and biological activity of 6-substituted 2-naphthoic acid retinoids.
Retinoic acid receptor beta,gamma-selective ligands: synthesis and biological activity of 6-substituted 2-naphthoic acid retinoids.
Journal of medicinal chemistry (1996-06-07)
K L Yu, P Spinazze, J Ostrowski, S J Currier, E J Pack, L Hammer, T Roalsvig, J A Honeyman, D R Tortolani, P R Reczek, M M Mansuri, J E Starrett
ABSTRACT
In search for retinoic acid receptor (RAR) selective ligands, a series of 6-substituted 2-naphthoic acid retinoids were synthesized and evaluated in vitro in a transactivation assay and a competition binding assay for all RARs. These derivatives, in general, showed RAR beta,gamma selectivity. Among these naphthoic acids, oxime derivative 12 was identified as a potent RAR gamma-selective retinoid, while olefinic derivative 11 was found to be comparable to retinoic acid and slightly RAR beta,gamma selective. For the bioassays, a general correlation was observed between the binding affinity of the ligand to the receptors and the potency of the compounds in the transactivation assay. The structure-activity relationship of these naphthoic acids will be discussed.
Sigma-Aldrich
2-Naphthoic acid, 98%