Merck
  • Cloning and characterization of KNR4, a yeast gene involved in (1,3)-beta-glucan synthesis.
Cloning and characterization of KNR4, a yeast gene involved in (1,3)-beta-glucan synthesis.
Molecular and cellular biology (1994-02-01)
Z Hong, P Mann, N H Brown, L E Tran, K J Shaw, R S Hare, B DiDomenico
ABSTRACT
k9 killer toxin from Hansenula mrakii was used to select a number of resistant mutants from Saccharomyces cerevisiae. Preliminary biochemical and genetic studies showed that some of them acquired structural defects in the cell wall. One of these mutants, the knr4-1 mutant, displays a number of cell wall defects, including osmotic sensitivity; sensitivity to cercosporamide, a known antifungal agent; and resistance to Zymolyase, a (1,3)-beta-glucanase. We report here the isolation and analysis of the KNR4 gene. DNA sequence analysis revealed an uninterrupted open reading frame which contains five potential start codons. The longest coding template encodes a protein of 505 amino acids with a calculated molecular mass of 57,044 Da. A data base search revealed 100% identity with a nuclear protein, SMI1p. Disruption of the KNR4 locus does not result in cell death; however, it leads to reduced levels of both (1,3)-beta-glucan synthase activity and (1,3)-beta-glucan content in the cell wall. The gene was mapped to the right arm of chromosome VII.
Sigma-Aldrich
Cercosporamide from Cercosporidium henningsii, ≥98% (HPLC)