Merck
Metabolism of steroid acetates by Streptomyces albus.
Journal of steroid biochemistry (1984-03-01)
A Mukherjee, S B Mahato
ABSTRACT
Fermentation of 16-dehydropregnenolone acetate (1a) with Streptomyces albus yielded 16-dehydropregnenolone (1b) and 16-dehydroprogesterone (IIa). Similar incubation of pregnenolone acetate (Ic) with the strain afforded pregnenolone (Id), progesterone (IIb) and 20 alpha-hydroxy progesterone (IIc) while dehydroepiandrosterone acetate (IIIa) under the conditions was converted to dehydroepiandrosterone (IIIb), androstenedione (IVa) and testosterone (IVc). The strain was also capable of converting testosterone acetate (IVb) having the 17-acetoxy function in the 5-membered D-ring to testosterone (IVc) and androstenedione (IVa). All the products were identified by the application of various chemical and spectrometric techniques.
Sigma-Aldrich
16-Dehydropregnenolone acetate, ≥95%