Merck
  • Tumoricidal effects of sodium hexachloroiridate on an ascitic tumor in mice.
Tumoricidal effects of sodium hexachloroiridate on an ascitic tumor in mice.
Cancer research (1983-07-01)
G M Kolodny, C Rose
ABSTRACT
Sodium hexachloroiridate injected 1 day after i.p. injection of 10(6) mouse ovarian tumor cells prevents the appearance of ascitic tumors in mice. Mice given injections of tumor cells all die at 20 to 30 days after tumor injection. Mice treated with sodium hexachloroiridate after tumor injection either have significant prolongation of life span without ascites or show no evidence of cancer even up to 7 months after tumor injection.
Sigma-Aldrich
Potassium hexachloroiridate(IV), 99.99% trace metals basis
Sigma-Aldrich
Potassium hexachloroiridate(IV), technical grade