Merck
  • Reactions of linoleic acid peroxyl radicals with phenolic antioxidants: a pulse radiolysis study.
Reactions of linoleic acid peroxyl radicals with phenolic antioxidants: a pulse radiolysis study.
International journal of radiation biology and related studies in physics, chemistry, and medicine (1987-09-01)
M Erben-Russ, W Bors, M Saran
ABSTRACT
Linoleic acid peroxyl radicals (LOO.) can be viewed as model intermediates occurring during lipid peroxidation processes. Formation and reactions of these species were investigated in aqueous alkaline solution using the technique of pulse radiolysis combined with kinetic spectroscopy. Irradiation of linoleic acid in N2O/O2-saturated solutions leads to a mixture of peroxyl radical isomers, whereas reaction of 13-hydroperoxylinoleic acid (13-LOOH) with azide radicals in N2O-saturated solution produces 13-LOO. radicals specifically. These peroxyl radicals cannot be observed directly, but their reactions with the two flavonols, kaempferol and quercetin, acting as radical-scavenging antioxidants, produced strongly absorbing aroxyl radicals (ArO.). The same aroxyl radicals were generated by .OH and N3. with rate constants exceeding 10(9) dm3 mol-1 s-1. Applying a reaction scheme that includes competing generation and decay reactions of both LOO. and ArO. radicals, we derived individual rate constants for LOO. reactions with the phenols (greater than 10(7) dm3 mol-1 s-1), with the aroxyl radicals to form covalent adducts (greater than 10(8) dm3 mol-1 s-1), as well as for their bimilecular decay (3.0 X 10(8) dm3 mol-1 s-1). These results demonstrate the high reactivity of both fatty acid peroxyl radicals and the flavone antioxidants in aqueous solution.
Sigma-Aldrich
2,3-Dihydroxynaphthalene, ≥98.0% (HPLC)