Merck
  • Inhibition of fibrinolytic system by liquoid (polyanetholesulfonate). A contributing factor in the liquoid-induced renal cortical necrosis.
Inhibition of fibrinolytic system by liquoid (polyanetholesulfonate). A contributing factor in the liquoid-induced renal cortical necrosis.
Haemostasis (1987-01-01)
N Gilboa, T J Ryan, R MacColl
ABSTRACT
Liquoid induces microvascular thrombosis and renal cortical necrosis in experimental animals. We hypothesized that thrombosis and renal cortical necrosis may, at least in part, result from the inhibition of the fibrinolytic system by liquoid. Effects of liquoid on plasminogen activation by rat kidney, purified human tissue plasminogen activator (TPA), urokinase, streptokinase, and on the amidolytic activities of TPA, urokinase, and plasmin were studied using chromogenic substrates and clot lysis. Liquoid had a strong inhibitory effect on the fibrinolytic system in vivo and in vitro. The inhibition was most effective at the plasminogen activation level, with activation by streptokinase being most susceptible. The demonstrated stoichiometric binding between liquoid and plasminogen, and to a lesser degree the direct inactivation of plasminogen activators and plasmin, is probably responsible for the reduction of plasminogen activation in circulation and in the kidneys.
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt, powder
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt, BioUltra